Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lanthanum strontium manganite

Catalog No. 70429610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-429-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-429-610G Supplier Sigma Aldrich Supplier No. 70429610G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

LSM-20 is a state of art electrode material for solid oxide fuel cells (SOFCs). LSM-20 possesses both ionic and electronic conductivity and belongs to the ABO3 perovskite oxide family.Lanthanum strontium manganite (LSM-20/GDC 10) is a ferromagnetic material, a Sr-doped LaMnO3 that is mainly used as a cathode material for solid oxide fuel cells (SOFCs). It also acts as a magnetic fluid, which can further be used as an inducer of tumor hyperthermia.

Specifications

Quantity 10 g
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.