Learn More
Sigma Aldrich Lanthanum strontium manganite
Catalog No. 70426110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
Catalog No. Quantity
70-426-110G 10 g
1 options
Catalog No. 70-426-110G Supplier Sigma Aldrich Supplier No. 70426110G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Lanthanum strontium manganite (LSM-35) is a ferromagnetic material, a Sr-doped LaMnO3 that is mainly used as a cathode material for solid oxide fuel cells (SOFCs).
Specifications
Quantity 10 g