Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthionine 98%, Thermo Scientific™

Catalog No. AC125102500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC125102500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC125102500 Supplier Thermo Scientific Chemicals Supplier No. 125102500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 922-55-4
Molecular Formula C6H12N2O4S
Molecular Weight (g/mol) 208.232
InChI Key DWPCPZJAHOETAG-WUCPZUCCSA-N
Synonym l,l-lanthionine, .beta.,.beta.'-thiodialanine, bis 2-amino-2-carboxyethyl sulfide, sulfide, bis 2-amino-2-carboxyethyl, alanine,3'-thiodi, alanine,3'-thiobis, l-cysteine, r, alanine,3'-thiodi-, l, sulfide, r-r*,r*, r-2-amino-3-2-amino-2-carboxyethyl thio propionic acid
PubChem CID 256406
IUPAC Name (2R)-2-amino-3-(2-amino-2-carboxyethyl)sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)SCC(C(=O)O)N

Specifications

Quantity 250 mg
Merck Index 15, 5416
Formula Weight 208.23
Chemical Name or Material Lanthionine, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.