Learn More
Lanx Chemical Protective Integrated Gloves
Catalog No. 191300238D
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-0238D X-Large
19-130-0238A Small
19-130-0238B Medium
19-130-0238C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-0238D Supplier Lanx Supplier No. WGXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Butyl rubber with TuffStuff leather palm for a more secure grip.
  • Cotton liner
  • Butyl rubber with leather palm
  • Ideal for chemical and biological applications
  • Vacuum packed
Specifications
Color Black
Size X-Large