Learn More
Lanx Chemical Protective Integrated Gloves
Description
- Cotton liner
- Butyl rubber with leather palm
- Ideal for chemical and biological applications
- Vacuum packed
Specifications
Specifications
| Color | Black |
| Size | Large |