Learn More
Key Scientific Products LAP Discs, with PEP Reagent, 50/pack
SDP
Catalog No. NC9979114
Encompass_Preferred
Used to detect the enzyme leucine aminopeptidase, one of the tests in the definitive identification of catalase negative gram positive cocci. Includes reagent and discs for 50 tests per jar.
Catalog No. NC9979114 Supplier Key Scientific Products Supplier No. K1378B
Used to detect the enzyme leucine aminopeptidase, one of the tests in the definitive identification of catalase negative gram positive cocci. Includes reagent and discs for 50 tests per jar.