Learn More
ACROBiosystems ACROBiosystems Human LAP (TGF-beta 1) Protein, His Tag
Supplier:  ACROBiosystems LAPH52451MG
Encompass_Preferred
Recombinant Protein;1MG;Human LAP (TGF-beta 1), His Tag (LAP-H5245) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (Accession # NP_000651 (C33S)).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 29.8 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAP (TGF-beta 1), His Tag (Cat. No. LAP-H5245) at 1 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-7337