Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LAP (TGF-beta 1) Protein, His,Avitag™
Catalog No. 501957116
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human LAP (TGF-beta 1), His,Avitag (LAP-H82Q6) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (Accession # P01137-1 (C33S)).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 32.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ITGAV&ITGB6 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT6-H52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LAP (TGF-beta 1), His,Avitag (Cat. No. LAP-H82Q6) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LAP (TGF-beta 1), His,Avitag (Cat. No. LAP-H82Q6) with a linear range of 0.4-6 ng/mL (Routinely tested).
Catalog No. 50-195-7116 Supplier ACROBiosystems Supplier No. LAPH82Q6200UG
Recombinant Protein;200UG;Biotinylated Human LAP (TGF-beta 1), His,Avitag (LAP-H82Q6) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (Accession # P01137-1 (C33S)).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 32.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ITGAV&ITGB6 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT6-H52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LAP (TGF-beta 1), His,Avitag (Cat. No. LAP-H82Q6) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LAP (TGF-beta 1), His,Avitag (Cat. No. LAP-H82Q6) with a linear range of 0.4-6 ng/mL (Routinely tested).