Learn More
Med Vet International Lap Sponge Sterile, 18" X 18", 40 Packs Of 5, 200/Case
Catalog No. 502831767
Encompass_Preferred
Laparotomy Sponges Pre-washed, 100% high quality cotton with tight weave Cross stitched with super locked edges to minimize linting and loose threads X-ray detectable Banded 5’sNot made with natural rubber latex
Catalog No. 50-283-1767 Supplier Med Vet International Supplier No. LAP1818CS
Laparotomy Sponges Pre-washed, 100% high quality cotton with tight weave Cross stitched with super locked edges to minimize linting and loose threads X-ray detectable Banded 5’sNot made with natural rubber latex