Learn More
Sino Biological LAP3 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 15480T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LAP3 (rh LAP3; Catalog#15480-H08H; NP_056991.2; Met1-Ala519). LAP3 specific IgG was purified by Human LAP3 affinity chromatography.
Catalog No. 50-224-8764