Learn More
Medchemexpress LLC HY-N6961 10mg Medchemexpress, Lapachol CAS:84-79-7 Purity:>98%
SDP
Catalog No. 501970484
Encompass_Preferred
Medchemexpress, HY-N6961 10mg Lapachol CAS:84-79-7 Lapachol is a naphthoquinone that was first isolated from Tabebuia avellanedae (Bignoniaceae). Lapachol shows anti-infection and antitumor activity Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0484 Supplier Medchemexpress LLC Supplier No. HYN696110MG
Medchemexpress, HY-N6961 10mg Lapachol CAS:84-79-7 Lapachol is a naphthoquinone that was first isolated from Tabebuia avellanedae (Bignoniaceae). Lapachol shows anti-infection and antitumor activity Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.