Learn More
Medchemexpress LLC Lapachol | 84-79-7 | 50 MG
SDP
Catalog No. 5000418384
Encompass_Preferred
Lapachol is a natural naphthoquinone that acts as an orally active, potent DHODH inhibitor. It exhibits immunosuppressive activity on lymphocytes by blocking DHODH activity and inhibiting pyrimidine synthesis. Lapachol is also a vitamin K antagonist with antitumor activity, capable of inhibiting DNA and RNA synthesis in neoplastic cells. Additionally, it possesses anti-Leishmania activity.
  • Potent DHODH inhibitor
  • Immunosuppressive activity on lymphocytes
  • Inhibits pyrimidine synthesis
  • Vitamin K antagonist
  • Antitumor activity
  • Inhibits DNA and RNA synthesis in neoplastic cells
  • Exhibits anti-Leishmania activity
  • Ameliorates experimental autoimmune arthritis
  • Reduces leukocyte infiltration in knee joints
Catalog No. 50-004-18384 Supplier Medchemexpress LLC Supplier No. HYN696150MG
Lapachol is a natural naphthoquinone that acts as an orally active, potent DHODH inhibitor. It exhibits immunosuppressive activity on lymphocytes by blocking DHODH activity and inhibiting pyrimidine synthesis. Lapachol is also a vitamin K antagonist with antitumor activity, capable of inhibiting DNA and RNA synthesis in neoplastic cells. Additionally, it possesses anti-Leishmania activity.
  • Potent DHODH inhibitor
  • Immunosuppressive activity on lymphocytes
  • Inhibits pyrimidine synthesis
  • Vitamin K antagonist
  • Antitumor activity
  • Inhibits DNA and RNA synthesis in neoplastic cells
  • Exhibits anti-Leishmania activity
  • Ameliorates experimental autoimmune arthritis
  • Reduces leukocyte infiltration in knee joints