Learn More
Cayman Chemical β-Lapachone, Purity: ≥98%, CAS: 4707-32-8, 10 mg
SDP
Catalog No. 502973021
Encompass_Preferred
A quinone which inhibits topoisomerase I at 25 μM and transglutaminase (IC50 = 5 μM); reduced to β-lapachone hydroquinone by NQO1, generating ROS during its reversion to β-lapachone; sensitizes cancer cells to radiation, suppresses NF-κB activation, and drives apoptosis
Catalog No. 50-297-3021 Supplier Cayman Chemical Supplier No. 1502110 MG
A quinone which inhibits topoisomerase I at 25 μM and transglutaminase (IC50 = 5 μM); reduced to β-lapachone hydroquinone by NQO1, generating ROS during its reversion to β-lapachone; sensitizes cancer cells to radiation, suppresses NF-κB activation, and drives apoptosis