Learn More
Medchemexpress LLC HY-13555 100mg Medchemexpress, β-Lapachone ARQ-501;NSC-26326;NSC-629749;SL-11001;beta-lapachone CAS:4707-32-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13555100MG
Encompass_Preferred
Medchemexpress,HY-13555 100mg β-Lapachone ARQ-501;NSC-26326;NSC-629749;SL-11001;beta-lapachone CAS:4707-32-8 Formula:C15H14O3 β-Lapachone is a naturally occurring O-naphthoquinone, acts as a topoisomerase I inhibitor, and induces apoptosis by inhibiting cell cycle progression. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7977