Learn More
Medchemexpress LLC Lapaquistat acetate | 189060-13-7 | 99.1% | 645.14 | 1 MG
SDP
Catalog No. 5000365233
Encompass_Preferred
Lapaquistat acetate (TAK-475) is a squalene synthase inhibitor that blocks the conversion of farnesyl diphosphate (FPP) to squalene in the cholesterol biosynthesis pathway. It is effective at lowering low-density lipoprotein cholesterol and is used for hypercholesterolemia and mevalonate kinase deficiency (MKD) research.
  • Squalene synthase inhibitor
  • Blocks conversion of farnesyl diphosphate (FPP) to squalene
  • Lowers low-density lipoprotein cholesterol
  • Used for hypercholesterolemia research
  • Used for mevalonate kinase deficiency (MKD) research
  • Solid appearance, white to off-white color
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-65233 Supplier Medchemexpress LLC Supplier No. 1MGHY16274
Lapaquistat acetate (TAK-475) is a squalene synthase inhibitor that blocks the conversion of farnesyl diphosphate (FPP) to squalene in the cholesterol biosynthesis pathway. It is effective at lowering low-density lipoprotein cholesterol and is used for hypercholesterolemia and mevalonate kinase deficiency (MKD) research.
  • Squalene synthase inhibitor
  • Blocks conversion of farnesyl diphosphate (FPP) to squalene
  • Lowers low-density lipoprotein cholesterol
  • Used for hypercholesterolemia research
  • Used for mevalonate kinase deficiency (MKD) research
  • Solid appearance, white to off-white color
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month