Learn More
Cayman Chemical Lapatinib, 231277-92-2, 10 mg
SDP
Supplier:  Cayman Chemical 1149310
Encompass_Preferred
A dual inhibitor of EGFR and ErbB2 (IC50s = 19 and 3 nM, respectively); inhibits the growth of EGFR-overexpressing A431 skin cancer and ErbB2-overexpressing SK-BR-3 breast cancer cells (IC50s = 0.14 and 0.124 μM, respectively); inhibits the growth of ErbB2-amplified OD19 esophageal and NCI-N87 gastric cancer cells (IC50s = 0.09 and 0.01 μM, respectively); inhibits the growth of gastric cancer cells in which ErbB2 is not amplified (IC50s = 0.35-8.58 μM); induces apoptosis in NCI-N87 and OD19 cells at 1 μM; reduces tumor growth in a BT474 breast cancer mouse xenograft model at 50 mg/kg; reduces tumor growth in an NCI-N87 mouse xenograft model at 100 mg/kg and induces tumor regression when used in combination with trastuzumab
Catalog No. NC0858287