Learn More
Selleck Chemical LLC Lapatinib (GW-572016) Ditosylate 10mM/1mL 388082-77-7
SDP
Catalog No. NC0981239
Encompass_Preferred
Lapatinib (GW-572016) Ditosylate is a potent EGFR and ErbB2 inhibitor with IC50 of 10.8 and 9.2 nM in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0981239 Supplier Selleck Chemical LLC Supplier No. S102810MM/1ML
Lapatinib (GW-572016) Ditosylate is a potent EGFR and ErbB2 inhibitor with IC50 of 10.8 and 9.2 nM in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.