Learn More
Medchemexpress LLC HY-50898 500mg , Lapatinib CAS:231277-92-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50898500MG
Encompass_Preferred
HY-50898 500mg Lapatinib CAS: 231277-92-2 Lapatinib (GW572016) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1085