Learn More
Medchemexpress LLC Lapatinib-d4 | 1184263-99-7 | 99.0% | 585.08 | C29H22D4ClFN4O4S | 10 MG
SDP
Catalog No. 5000298482
Encompass_Preferred
Lapatinib-d4 is the deuterium-labeled isotopologue of lapatinib used as a stable isotope standard for analytical and pharmacokinetic studies. It is supplied as a solid powder in pre-weighed quantities with specified storage recommendations for long-term stability.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High purity (≈99%) suitable for quantitative analysis.
  • Offered in small pre-weighed amounts for method development.
  • Stable in powder form under low-temperature storage.
  • Comes with a product datasheet and safety data sheet for handling.
Catalog No. 50-002-98482 Supplier Medchemexpress LLC Supplier No. HY50898S10MG
Lapatinib-d4 is the deuterium-labeled isotopologue of lapatinib used as a stable isotope standard for analytical and pharmacokinetic studies. It is supplied as a solid powder in pre-weighed quantities with specified storage recommendations for long-term stability.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • High purity (≈99%) suitable for quantitative analysis.
  • Offered in small pre-weighed amounts for method development.
  • Stable in powder form under low-temperature storage.
  • Comes with a product datasheet and safety data sheet for handling.