Learn More
Medchemexpress LLC Lapatinib-d4 (GW572016-d4; GW2016-d4) | 1184263-99-7 | 99.0% | C29H22D4ClFN4O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY50898S5MG
Encompass_Preferred
Lapatinib-d4 is the deuterium-labeled version of Lapatinib, a potent, orally active inhibitor of the ErbB-2 and EGFR tyrosine kinase domains. It is suitable for research use only. Deuteration, which involves incorporating stable heavy isotopes into drug molecules, serves as a tracer for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-58867