Learn More
Cayman Chemical LapatInIb-d4tosylate 500ug
SDP
Catalog No. 502935571
Encompass_Preferred
An internal standard for the quantification of lapatinib by GC- or LC-MS
Catalog No. 50-293-5571 Supplier Cayman Chemical Supplier No. 25459500 UG
An internal standard for the quantification of lapatinib by GC- or LC-MS