Learn More
Medchemexpress LLC HY-50898A 100mg , Lapatinib (ditosylate) CAS:388082-77-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50898A100MG
Encompass_Preferred
HY-50898A 100mg Lapatinib (ditosylate) CAS: 388082-77-7 Lapatinib ditosylate (GW572016 ditosylate) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0257