Learn More
Medchemexpress LLC HY-50898A 1g , Lapatinib (ditosylate) GW-572016 ditosylate CAS:388082-77-7 Purity:98%
SDP
Catalog No. 501492295
Encompass_Preferred
Medchemexpress, HY-50898A 1g Lapatinib (ditosylate) GW-572016 ditosylate CAS:388082-77-7 Formula:C43H42ClFN4O10S3 IC50: 10.2 nM (EGFR), 9.8 nM (ErbB2) Purity:98% Lapatinib ditosylate is a potent EGFR and ErbB2 inhibitor with IC 50 of 10.2 and 9.8 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2295 Supplier Medchemexpress LLC Supplier No. HY50898A1G
Medchemexpress, HY-50898A 1g Lapatinib (ditosylate) GW-572016 ditosylate CAS:388082-77-7 Formula:C43H42ClFN4O10S3 IC50: 10.2 nM (EGFR), 9.8 nM (ErbB2) Purity:98% Lapatinib ditosylate is a potent EGFR and ErbB2 inhibitor with IC 50 of 10.2 and 9.8 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.