Learn More
STEMCELL Technologies Lapatinib​, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 73242
Encompass
Lapatinib is a selective dual inhibitor of epidermal growth factor receptor (EGFR, ERBB1) and human epidermal growth factor receptor 2 (HER2, ERBB2), each with an IC₅₀ of about 10 nM (Rusnak et al.). It binds reversibly, but with a very slow off-rate, to the ATP binding pocket in an inactive-like conformation, thus preventing autophosphorylation. Binding is very potent, with Ki values of 3 and 13 nM to EGFR and HER2 respectively (Wood et al.).CANCER RESEARCH· Induces apoptosis in cells overexpressing EGFR and/or HER2, and inhibits tumor growth in a mouse xenograft model (Xia et al.).
Catalog No. 50-197-6045