Learn More
Selleck Chemical LLC Lapatinib ditosylate monohydrate S5241-25mg
SDP
Catalog No. 503110952
Encompass_Preferred
Lapatinib ditosylate monohydrate (Lapatinib ditoluenesulfonate monohydrate Lapatinib tosilate Lapatinib tosilate hydrate)is a dual tyrosine kinase inhibitor which interrupts the HER2/neu and epidermal growth factor receptor (EGFR) pathways used for treatment of solid tumours such as breast and lung cancer
Catalog No. 50-311-0952 Supplier Selleck Chemical LLC Supplier No. S524125MG
Lapatinib ditosylate monohydrate (Lapatinib ditoluenesulfonate monohydrate Lapatinib tosilate Lapatinib tosilate hydrate)is a dual tyrosine kinase inhibitor which interrupts the HER2/neu and epidermal growth factor receptor (EGFR) pathways used for treatment of solid tumours such as breast and lung cancer