Learn More
Cayman Chemical LappaconItIn 100mg
SDP
Catalog No. 502938327
Encompass_Preferred
A diterpene alkaloid with diverse biological properties; an inhibitor of Nav1.7 (IC50 = 27.67 µM); Lappaconitine has antiarrhythmic activity in a rat model of aconitine-induced arrhythmia (ED50 = 0.05 mg/kg); increases the mechanical and thermal paw withdrawal thresholds in rats in a model of spinal nerve ligation-induced neuropathic pain (ED50s = 1.1 and 1.6 mg/kg, respectively)
Catalog No. 50-293-8327 Supplier Cayman Chemical Supplier No. 28747100 MG
A diterpene alkaloid with diverse biological properties; an inhibitor of Nav1.7 (IC50 = 27.67 µM); Lappaconitine has antiarrhythmic activity in a rat model of aconitine-induced arrhythmia (ED50 = 0.05 mg/kg); increases the mechanical and thermal paw withdrawal thresholds in rats in a model of spinal nerve ligation-induced neuropathic pain (ED50s = 1.1 and 1.6 mg/kg, respectively)