Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lappaol A | 62333-08-8 | 1 MG
SDP

Catalog No. 5000421106
Encompass_Preferred

Lappaol A is a natural compound exhibiting antiaging activity, primarily intended for research use. This off-white to light yellow solid has a high purity of 98.63% and a molecular weight of 536.57.

  • Exhibits antiaging activity
  • Purity: 98.63%
  • Molecular weight: 536.57
  • Formula: C30H32O9
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Structure classification: Phenylpropanoids, Lignans, Phenols, Polyphenols
  • Derived from plants including Leguminosae and Compositae

Catalog No. 50-004-21106 Supplier Medchemexpress LLC Supplier No. HYN24751MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lappaol A is a natural compound exhibiting antiaging activity, primarily intended for research use. This off-white to light yellow solid has a high purity of 98.63% and a molecular weight of 536.57.

  • Exhibits antiaging activity
  • Purity: 98.63%
  • Molecular weight: 536.57
  • Formula: C30H32O9
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Structure classification: Phenylpropanoids, Lignans, Phenols, Polyphenols
  • Derived from plants including Leguminosae and Compositae

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.