Learn More
World Precision Instrument Lapping Film, Alumina, 0.3 micron
SDP
Catalog No. 502521461
Encompass
  • For use with SYS-48000 microbeveler.
Catalog No. 50-252-1461 Supplier World Precision Instrument Supplier No. 4801503
  • For use with SYS-48000 microbeveler.