Learn More
Aviva Systems Biology Corp LAPTM4A Antibody - middle region (ARP47057_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47057_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LAPTM4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LAPTM4A. Alternate names: HUMORF13, KIAA0108, LAPTM4, MBNT, MtrpApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59461