Learn More
Selleck Chemical LLC Dacinostat (LAQ824) 50mg 404951-53-7 NVP-LAQ824
SDP
Supplier:  Selleck Chemical LLC S109550MG
Encompass_Preferred
Dacinostat (LAQ824, NVP-LAQ824) is a novel HDAC inhibitor with IC50 of 32 nM and is known to activate the p21 promoter. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-873-0