Learn More
Apexbio Technology LLC Laquinimod (ABR-215062) 248281-84-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061715
Encompass_Preferred
Laquinimod (ABR-215062 CAS 248281-84-7) is an orally bioavailable immunomodulatory small molecule Research studies demonstrate that in experimental autoimmune encephalomyelitis (EAE) a widely utilized rodent model for studying human multiple sclerosis (MS) laquinimod treatment suppresses disease progression in a dose-dependent manner Mechanistically the compound reduces inflammatory infiltration of CD4 T lymphocytes and macrophages into the central nervous system thereby modulating the Th1/Th2 immune response and inducing regulatory cytokines such as TGF- Clinical investigations into relapsing MS indicate that laquinimod administration correlates with reduced disease activity clinical relapse rate MRI-measured lesions and brain volume loss
Catalog No. 50-000-61715 Supplier Apexbio Technology LLC Supplier No. A84595.1
Laquinimod (ABR-215062 CAS 248281-84-7) is an orally bioavailable immunomodulatory small molecule Research studies demonstrate that in experimental autoimmune encephalomyelitis (EAE) a widely utilized rodent model for studying human multiple sclerosis (MS) laquinimod treatment suppresses disease progression in a dose-dependent manner Mechanistically the compound reduces inflammatory infiltration of CD4 T lymphocytes and macrophages into the central nervous system thereby modulating the Th1/Th2 immune response and inducing regulatory cytokines such as TGF- Clinical investigations into relapsing MS indicate that laquinimod administration correlates with reduced disease activity clinical relapse rate MRI-measured lesions and brain volume loss