Learn More
Apexbio Technology LLC Laquinimod (ABR-215062), 5mg. Cas: 248281-84-7 MFCD: MFCD08689859. Immunomodulator,orally-available,quinolinone-based and broad spectrum
SDP
Supplier:  Apexbio Technology LLC A84595
Encompass_Preferred
ABR-215062 is an orally active immunoregulator. Experimental autoimmune encephalomyelitis (EAE) is an inflammatory autoimmune disease of the CNS that can be elicited in rodents and represents the major animal model for the study of multiple sclerosis (MS). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5147