Learn More
Medchemexpress LLC Laquinimod-d5 | 1214267-09-0 | 98.3% | 1 MG
SDP
Catalog No. 5000338949
Encompass_Preferred
Laquinimod-d5 is a deuterium labeled form of Laquinimod, an orally available carboxamide derivative. It acts as a potent immunomodulator, preventing neurodegeneration and inflammation in the central nervous system. This compound reduces astrocytic NF-κB activation, offering protection from Cuprizone-induced demyelination.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential for relapsing-remitting and chronic progressive forms of multiple sclerosis research.
  • Potential for neurodegenerative diseases research.
Catalog No. 50-003-38949 Supplier Medchemexpress LLC Supplier No. HY13010S1MG
Laquinimod-d5 is a deuterium labeled form of Laquinimod, an orally available carboxamide derivative. It acts as a potent immunomodulator, preventing neurodegeneration and inflammation in the central nervous system. This compound reduces astrocytic NF-κB activation, offering protection from Cuprizone-induced demyelination.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential for relapsing-remitting and chronic progressive forms of multiple sclerosis research.
  • Potential for neurodegenerative diseases research.