Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Laquinimod-d5 | 1214267-09-0 | 98.3% | 361.83 g/mol | C19H12D5ClN2O3 | 5 MG
SDP

Catalog No. 5000262860
Encompass_Preferred

Laquinimod-d5 is a deuterium-labeled analogue of laquinimod provided for research use as an analytical reference and internal standard. It is intended for isotope-labeling studies, quantitative mass spectrometry, and method development in neurodegeneration and immunomodulation research. Supplied as a high-purity powder, it supports accurate tracing and quantitation in analytical workflows.

  • Deuterium-labeled internal standard for mass spectrometry quantitation.
  • High purity (98.3%) suitable for analytical applications.
  • Consistent molecular composition (C19H12D5ClN2O3) for reproducible results.
  • Available in small milligram quantities for method development.
  • Recommended powder storage at -20°C to preserve stability.

Catalog No. 50-002-62860 Supplier Medchemexpress LLC Supplier No. HY13010S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laquinimod-d5 is a deuterium-labeled analogue of laquinimod provided for research use as an analytical reference and internal standard. It is intended for isotope-labeling studies, quantitative mass spectrometry, and method development in neurodegeneration and immunomodulation research. Supplied as a high-purity powder, it supports accurate tracing and quantitation in analytical workflows.

  • Deuterium-labeled internal standard for mass spectrometry quantitation.
  • High purity (98.3%) suitable for analytical applications.
  • Consistent molecular composition (C19H12D5ClN2O3) for reproducible results.
  • Available in small milligram quantities for method development.
  • Recommended powder storage at -20°C to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.