Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Laquinimod-d5 | 1214267-09-0 | MFCD08689859 | 98.3% | C19H12D5ClN2O3 | 10MG
SDP

Catalog No. 5000260007
Encompass_Preferred

Laquinimod-d5 is a deuterium-labeled stable isotope of laquinimod intended for use as an analytical reference standard. It is primarily applied as an internal standard in mass spectrometry and for ADME/metabolism and quantitative studies where isotopic labeling improves tracking and accuracy. The material is characterized and supplied with supporting analytical data.

  • Deuterium-labeled (d5) isotopologue for mass spectrometry internal standards.
  • High purity for analytical use (98.3%).
  • Suitable for ADME, metabolism, and quantitative analysis.
  • Molecular formula C19H12D5ClN2O3; molecular weight 361.83.
  • Supplied with certificate of analysis and supporting characterization.
  • Facilitates accurate tracing and quantification in bioanalytical workflows.

Catalog No. 50-002-60007 Supplier Medchemexpress LLC Supplier No. HY13010S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laquinimod-d5 is a deuterium-labeled stable isotope of laquinimod intended for use as an analytical reference standard. It is primarily applied as an internal standard in mass spectrometry and for ADME/metabolism and quantitative studies where isotopic labeling improves tracking and accuracy. The material is characterized and supplied with supporting analytical data.

  • Deuterium-labeled (d5) isotopologue for mass spectrometry internal standards.
  • High purity for analytical use (98.3%).
  • Suitable for ADME, metabolism, and quantitative analysis.
  • Molecular formula C19H12D5ClN2O3; molecular weight 361.83.
  • Supplied with certificate of analysis and supporting characterization.
  • Facilitates accurate tracing and quantification in bioanalytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.