Learn More
TARGETMOL CHEMICALS INC Larazotide acetate 10MG
SDP
Catalog No. 502878532
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Larazotide acetate is a synthetic peptide. It functions as a tight junction regulator and reverses leaky junctions to their normally closed state. Purity 95.53%
Catalog No. 50-287-8532 Supplier TARGETMOL CHEMICALS INC Supplier No. T1571110MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Larazotide acetate is a synthetic peptide. It functions as a tight junction regulator and reverses leaky junctions to their normally closed state. Purity 95.53%