Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Larazotide acetate | 881851-50-9 | MFCD31630844 | ≥98.0% | 785.89 g/mol | C34H59N9O12 | 25 MG
SDP

Catalog No. 5000243743
Encompass_Preferred

Larazotide acetate is an eight-amino-acid synthetic peptide zonulin antagonist used in research to modulate tight junctions and intestinal permeability. Supplied as a lyophilized acetate salt, the compound is intended for laboratory studies and requires sealed, low-temperature storage to maintain stability.

  • Sequence: Gly-Gly-Val-Leu-Val-Gln-Pro-Gly.
  • High purity specification (≥98.0% by HPLC) suitable for analytical and preclinical studies.
  • Molecular weight available for dosing calculations: 785.89 g/mol.
  • Stable as a dry powder when stored sealed at -20°C to -80°C.
  • Provided as a lyophilized powder for convenient reconstitution.
  • Intended for research use only; not for clinical or diagnostic use.

Catalog No. 50-002-43743 Supplier Medchemexpress LLC Supplier No. HY106268A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Larazotide acetate is an eight-amino-acid synthetic peptide zonulin antagonist used in research to modulate tight junctions and intestinal permeability. Supplied as a lyophilized acetate salt, the compound is intended for laboratory studies and requires sealed, low-temperature storage to maintain stability.

  • Sequence: Gly-Gly-Val-Leu-Val-Gln-Pro-Gly.
  • High purity specification (≥98.0% by HPLC) suitable for analytical and preclinical studies.
  • Molecular weight available for dosing calculations: 785.89 g/mol.
  • Stable as a dry powder when stored sealed at -20°C to -80°C.
  • Provided as a lyophilized powder for convenient reconstitution.
  • Intended for research use only; not for clinical or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.