Learn More
Medchemexpress LLC Larazotide acetate | 881851-50-9 | MFCD31630844 | 99.7% | 785.89 g/mol | C34H59N9O12 | 1 MG
SDP
Catalog No. 5000244376
Encompass_Preferred
Larazotide acetate is a synthetic peptide zonulin antagonist used in preclinical research to modulate intestinal tight junction permeability. Supplied as a solid powder for laboratory research, it has documented activity in cell-based and animal models affecting barrier function.
  • Orally active zonulin antagonist that modulates tight junctions.
  • Characterized in cell-based assays and animal models for reducing intestinal permeability.
  • High purity (99.68%) suitable for research applications.
  • Solid, white to off-white powder for laboratory use.
  • Storage: powder stable at -80°C for up to 2 years and -20°C for up to 1 year.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-44376 Supplier Medchemexpress LLC Supplier No. HY106268A1MG
Add to Cart
Add to Cart
Larazotide acetate is a synthetic peptide zonulin antagonist used in preclinical research to modulate intestinal tight junction permeability. Supplied as a solid powder for laboratory research, it has documented activity in cell-based and animal models affecting barrier function.
  • Orally active zonulin antagonist that modulates tight junctions.
  • Characterized in cell-based assays and animal models for reducing intestinal permeability.
  • High purity (99.68%) suitable for research applications.
  • Solid, white to off-white powder for laboratory use.
  • Storage: powder stable at -80°C for up to 2 years and -20°C for up to 1 year.
  • Intended for research use only; not for human or clinical use.