Learn More
Medchemexpress LLC Larazotide (acetate) | 881851-50-9 | MFCD31630844 | 99.7% | 785.89 g/mol | C34H59N9O12 | 100 MG
SDP
Catalog No. 5000243745
Encompass_Preferred
Larazotide acetate is an eight-amino-acid peptide zonulin antagonist supplied as a research reagent. It is used in studies of celiac disease and intestinal barrier dysfunction and has reported antiviral activity against varicella-zoster virus. For research use only; not for human therapeutic use.
  • Eight-amino-acid peptide sequence: Gly-Gly-Val-Leu-Val-Gln-Pro-Gly.
  • Orally active zonulin antagonist that modulates tight junctions.
  • Reported in vitro antiviral activity versus varicella-zoster virus.
  • Supplied as a powder; recommended sealed, cold storage to preserve stability.
  • Well characterized molecular weight and formula for research applications.
Catalog No. 50-002-43745 Supplier Medchemexpress LLC Supplier No. HY106268A100MG
Larazotide acetate is an eight-amino-acid peptide zonulin antagonist supplied as a research reagent. It is used in studies of celiac disease and intestinal barrier dysfunction and has reported antiviral activity against varicella-zoster virus. For research use only; not for human therapeutic use.
  • Eight-amino-acid peptide sequence: Gly-Gly-Val-Leu-Val-Gln-Pro-Gly.
  • Orally active zonulin antagonist that modulates tight junctions.
  • Reported in vitro antiviral activity versus varicella-zoster virus.
  • Supplied as a powder; recommended sealed, cold storage to preserve stability.
  • Well characterized molecular weight and formula for research applications.