Learn More
LabScientific Large, insulated, rectangluar ice pan with lid (minimal evaporation). Precise pour spout. Handles. Stackable. Low temperature to -80°C.
Catalog No. NC9382684
Encompass
Rectangular Ice Pan and Test Tube Rack; 9L; Insulated; Red; Large; Four angled sections allow for ice and plenty of room for organization of .1mL to 25mL vials, tubes, pipets or pipet cannisters; Interior dimensions 38.0x23.0x13.5 cm; exterior dimensions 52.1 x 29.2 x 16.3 cm
Catalog No. NC9382684 Supplier LabScientific Supplier No. RECB1201
Rectangular Ice Pan and Test Tube Rack; 9L; Insulated; Red; Large; Four angled sections allow for ice and plenty of room for organization of .1mL to 25mL vials, tubes, pipets or pipet cannisters; Interior dimensions 38.0x23.0x13.5 cm; exterior dimensions 52.1 x 29.2 x 16.3 cm