Learn More
OHAUS Large Ampule/Tube Attachment
Catalog No. 01804406 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-406 Vortex Attachment
1 options
Catalog No. 01-804-406 Supplier OHAUS Supplier No. 83041391
Add to Cart
Add to Cart
Ideal for use with the Vortex Genie 2 family of mixers and the Vortex Genie Pulse.
  • Provides vigorous end-to-end agitation combined with vortexing.
  • Handles up to four 10 mm to 17 mm diameter tubes or ampules.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse