Learn More
Stoelting™ Large Animal Adaptor
SDP
Catalog No. 10000153 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Cat
Model:
For 51600
Non Puncture
Non Rupture
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Model
10-000-153 Cat For 51600
10-000-154 Cat Non Rupture
10-000-223 Cat Non Puncture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-000-153 Supplier Stoelting™ Supplier No. 51626
Only null left
Add to Cart
Add to Cart
Accommodate a wide size range of large animals in either the Lab Standard™ or Parallel Rail series of stereotaxic instruments
Dog Adaptor available only for the Parallel Rail (51800) series stereotaxic instruments. To fit any size dog, it should be used with 51850 and 51851 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.
Cat Adaptor is available for the Lab Standard™ or Parallel Rail series of stereotaxic instruments. The adaptor for the Lab Standard™ series (51600) includes spacers and bolts to raise the U frame 2.5 inches above the base plate, the eye socket/palate holder and 18° ear bars. Also available with  non-puncture 45° ear bars (51827).
Specifications
For Use With (Application) Neuroscience
Animal Cat
Model For 51600