Learn More
World Precision Instrument Instrument Rodent Guillotine, For Larger Animals, 4.0 x 4.0in Opening
SDP
Catalog No. 50822918
Encompass
These guillotines are designed for for larger animals. The guillotine is considered one of the most humane methods of dispatching laboratory animals. With a single downward motion of the handle, you can euthanize a rodent quickly and without trauma.
Catalog No. 50-822-918 Supplier World Precision Instrument Supplier No. DCAPL
These guillotines are designed for for larger animals. The guillotine is considered one of the most humane methods of dispatching laboratory animals. With a single downward motion of the handle, you can euthanize a rodent quickly and without trauma.