Learn More
Med Vet International Large Animal IV Catheter, Sterile, 10g x 5.5in, J0457B
Catalog No. 5000581748
Encompass_Preferred
Large animals need large volumes of IV fluids in a short period of time. Naturally, the IV catheter needs to be a large bore, and often is even put in bilaterally to maximize fluid administration speed.Features:Large bore: 10g and 14g catheters. The 10g is the largest available.Longer lengths: 51/2" catheter lessens the risk of pulling out the catheter.Pebax catheter implant grade material: much less bioreactive to lessen thrombosis formation for long term >3 days placement.Kink resistant: other catheters like Teflon will kink upon bending, making them unusable.Clear catheter material versus opaque Teflon.Suture patch: a thin, flexible patch with twin eyelets. Allows suturing in place or hold with tissue adhesives. Reduces catheter pull-out.Competitive pricing and available through JorVet distributors.
Catalog No. 50-005-81748 Supplier Med Vet International Supplier No. JOR457B
Large animals need large volumes of IV fluids in a short period of time. Naturally, the IV catheter needs to be a large bore, and often is even put in bilaterally to maximize fluid administration speed.Features:Large bore: 10g and 14g catheters. The 10g is the largest available.Longer lengths: 51/2" catheter lessens the risk of pulling out the catheter.Pebax catheter implant grade material: much less bioreactive to lessen thrombosis formation for long term >3 days placement.Kink resistant: other catheters like Teflon will kink upon bending, making them unusable.Clear catheter material versus opaque Teflon.Suture patch: a thin, flexible patch with twin eyelets. Allows suturing in place or hold with tissue adhesives. Reduces catheter pull-out.Competitive pricing and available through JorVet distributors.