Learn More
World Precision Instrument Large Animal Manipulator, Left handed
SDP
Catalog No. 502539118
Encompass
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Catalog No. 50-253-9118 Supplier World Precision Instrument Supplier No. 505328
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.