Learn More
World Precision Instrument Large Animal Manipulator, Right handed
SDP
Catalog No. 502539117
Encompass
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Catalog No. 50-253-9117 Supplier World Precision Instrument Supplier No. 505329
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.