Learn More
World Precision Instrument Large Animal Parallel Rail Stereotaxic, Base and Frame Only
SDP
Catalog No. 502539109
Encompass
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Reliable Locks
If it is necessary to tilt the stereotaxic frame to gain access to the animal’s side or underside, the tilting and angling universal joint on WPI Rat Stereotaxic Instruments manipulators have the most secure, reliable locks of any stereotaxic instrument.
Catalog No. 50-253-9109 Supplier World Precision Instrument Supplier No. 505342
Add to Cart
Add to Cart
Request a Quote
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Reliable Locks
If it is necessary to tilt the stereotaxic frame to gain access to the animal’s side or underside, the tilting and angling universal joint on WPI Rat Stereotaxic Instruments manipulators have the most secure, reliable locks of any stereotaxic instrument.