Learn More
World Precision Instrument Large Animal Stereotaxic, 2 Manipulator Arms
SDP
Catalog No. 502539120
Encompass
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Catalog No. 50-253-9120 Supplier World Precision Instrument Supplier No. 505326
Add to Cart
Add to Cart
Request a Quote
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.