Learn More
World Precision Instrument Large Animal Stereotaxic, Base and Frame Only
SDP
Catalog No. 502539119
Encompass
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Catalog No. 50-253-9119 Supplier World Precision Instrument Supplier No. 505327
Add to Cart
Add to Cart
Request a Quote
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.