Learn More
Doccol Corporation Large B MCAO suture 70L910
SDP
Catalog No. NC3243379
Encompass
Key Features:1. Filament Specifications: The 7-0 filament size has a diameter ranging from 0.06 to 0.09 mm and a length of 20 mm, providing balanced excellence in both the needed flexibility and pushability for working on the delicated vasculature.2. Silicone Rubber Coating: With a diameter of 0.22 +/- 0.01 mm, the silicone rubber coating ensures smooth and controlled insertion, enhancing precision in your experiments. Such coating provides ideal dimensions for MCAO procedures in mice within the body weight range of 25 +/- 2 grams. The coating extends 9-10 mm, catering to the needs of permanent MCAO or transient (less than 60 min) MCAO followed by reperfusion.3. Suitable for Highly Skilled Surgeons and for MCAO models that requires the highest consistence of infarct volume: The 9-10 mm coating length is optimized for providing the highest consistence and reliability of infarct volume.4. Reusable Design: Our silicone rubber-coate
Catalog No. NC3243379 Supplier Doccol Corporation Supplier No. 7022910PK5RE
Key Features:1. Filament Specifications: The 7-0 filament size has a diameter ranging from 0.06 to 0.09 mm and a length of 20 mm, providing balanced excellence in both the needed flexibility and pushability for working on the delicated vasculature.2. Silicone Rubber Coating: With a diameter of 0.22 +/- 0.01 mm, the silicone rubber coating ensures smooth and controlled insertion, enhancing precision in your experiments. Such coating provides ideal dimensions for MCAO procedures in mice within the body weight range of 25 +/- 2 grams. The coating extends 9-10 mm, catering to the needs of permanent MCAO or transient (less than 60 min) MCAO followed by reperfusion.3. Suitable for Highly Skilled Surgeons and for MCAO models that requires the highest consistence of infarct volume: The 9-10 mm coating length is optimized for providing the highest consistence and reliability of infarct volume.4. Reusable Design: Our silicone rubber-coate